Learn More
Enzo Life Sciences 8-CPT-cAMP, (20mg), CAS Number: 93882-12-3
SDP
Supplier:  Enzo Life Sciences BMLCN1300020
Encompass_Preferred
PKA activator. A lipophilic, cell-permeable cAMP analog which activates cAMP- and cGMP-dependent protein kinase.  More potent than cAMP and more resistant to phosphodiesterasese, with good selectivity for site B of PKA. For specific activation of the cAMP pathway the highly membrane permeable Sp-5,6-DCI-cBIMPS (Prod. No. BML-CN135). Formula: C16H14CIN5O6PS. Na. MW: 493.8. CAS Number: 93882-12-3. Purity: ≥96% (HPLC). Appearance: White solid. Solubility: Soluble in water (25mg/ml). Use/Stability: Store as supplied (solid) at -20°C for 1 year. Once dissolved in water, it can be stored frozen at -20°C up to 3 months. Long Term Storage: -20°C
Catalog No. NC9965560