Learn More
Medchemexpress LLC 8-CPT-Cyclic AMP sodium | 93882-12-3 | 99.7% | 493.79 | 10 MG
SDP
Catalog No. 5000439960
Encompass_Preferred
8-CPT-Cyclic AMP sodium is a selective activator of cyclic AMP-dependent protein kinase (PKA). It is also a potent inhibitor of the cyclic GMP-specific phosphodiesterase (PDE VA) with an IC50 of 0.9 μM. Additionally, 8-CPT-Cyclic AMP sodium inhibits PDE III and PDE IV with IC50 values of 5.5 μM and 6.6 μM, respectively, and is a potent Epac activator. It has been shown to delay TNF-α/CHX-induced and α-Fas-induced apoptosis in neutrophils.
  • Selective activator of cyclic AMP-dependent protein kinase (PKA).
  • Potent inhibitor of cyclic GMP-specific phosphodiesterase (PDE VA) with an IC50 of 0.9 μM.
  • Inhibits PDE III and PDE IV with IC50 values of 5.5 μM and 6.6 μM.
  • Potent Epac activator.
  • Delays TNF-α/CHX-induced and α-Fas-induced apoptosis in neutrophils.
Catalog No. 50-004-39960 Supplier Medchemexpress LLC Supplier No. HY11167310MG
8-CPT-Cyclic AMP sodium is a selective activator of cyclic AMP-dependent protein kinase (PKA). It is also a potent inhibitor of the cyclic GMP-specific phosphodiesterase (PDE VA) with an IC50 of 0.9 μM. Additionally, 8-CPT-Cyclic AMP sodium inhibits PDE III and PDE IV with IC50 values of 5.5 μM and 6.6 μM, respectively, and is a potent Epac activator. It has been shown to delay TNF-α/CHX-induced and α-Fas-induced apoptosis in neutrophils.
  • Selective activator of cyclic AMP-dependent protein kinase (PKA).
  • Potent inhibitor of cyclic GMP-specific phosphodiesterase (PDE VA) with an IC50 of 0.9 μM.
  • Inhibits PDE III and PDE IV with IC50 values of 5.5 μM and 6.6 μM.
  • Potent Epac activator.
  • Delays TNF-α/CHX-induced and α-Fas-induced apoptosis in neutrophils.