Learn More
Medchemexpress LLC 8-CPT-Cyclic AMP sodium | 93882-12-3 | 99.7% | 493.79 | 100 MG
SDP
Catalog No. 5000439136
Encompass_Preferred
8-CPT-Cyclic AMP sodium is a selective activator of cyclic AMP-dependent protein kinase (PKA). It is also a potent inhibitor of the cyclic GMP-specific phosphodiesterase (PDE VA) with an IC50 of 0.9 μM, and it inhibits PDE III and PDE IV with IC50Epac, functioning as a potent Epac activator.
  • Delays TNF-α/CHX-induced and α-Fas-induced apoptosis in neutrophils.
  • More effective against apoptosis induced by TNF-α/CHX than by α-Fas.
Catalog No. 50-004-39136 Supplier Medchemexpress LLC Supplier No. HY111673100MG
8-CPT-Cyclic AMP sodium is a selective activator of cyclic AMP-dependent protein kinase (PKA). It is also a potent inhibitor of the cyclic GMP-specific phosphodiesterase (PDE VA) with an IC50 of 0.9 μM, and it inhibits PDE III and PDE IV with IC50Epac, functioning as a potent Epac activator.
  • Delays TNF-α/CHX-induced and α-Fas-induced apoptosis in neutrophils.
  • More effective against apoptosis induced by TNF-α/CHX than by α-Fas.