Learn More
Medchemexpress LLC 8-CPT-Cyclic AMP sodium | 93882-12-3 | 99.7% | 493.79 | 25 MG
SDP
Catalog No. 5000439707
Encompass_Preferred
8-CPT-Cyclic AMP (8-CPT-cAMP) sodium is a selective activator of cyclic AMP-dependent protein kinase (PKA). It is also identified as a potent inhibitor of the cyclic GMP-specific phosphodiesterase (PDE VA) with an IC50 of 0.9 μM. Additionally, 8-CPT-Cyclic AMP sodium inhibits PDE III and PDE IV with IC50Epac and functions as a potent Epac activator.
  • Selective activator of cyclic AMP-dependent protein kinase (PKA).
  • Potent inhibitor of the cyclic GMP-specific phosphodiesterase (PDE VA) with an IC50 of 0.9 μM.
  • Inhibits PDE III and PDE IV.
  • Potent Epac activator.
  • Delays TNF-/CHX-induced and α-Fas-induced apoptosis in neutrophils.
Catalog No. 50-004-39707 Supplier Medchemexpress LLC Supplier No. 25MGHY111673
8-CPT-Cyclic AMP (8-CPT-cAMP) sodium is a selective activator of cyclic AMP-dependent protein kinase (PKA). It is also identified as a potent inhibitor of the cyclic GMP-specific phosphodiesterase (PDE VA) with an IC50 of 0.9 μM. Additionally, 8-CPT-Cyclic AMP sodium inhibits PDE III and PDE IV with IC50Epac and functions as a potent Epac activator.
  • Selective activator of cyclic AMP-dependent protein kinase (PKA).
  • Potent inhibitor of the cyclic GMP-specific phosphodiesterase (PDE VA) with an IC50 of 0.9 μM.
  • Inhibits PDE III and PDE IV.
  • Potent Epac activator.
  • Delays TNF-/CHX-induced and α-Fas-induced apoptosis in neutrophils.