Learn More
Medchemexpress LLC 8-CPT-Cyclic AMP sodium | 93882-12-3 | 99.7% | 493.79 | 50 MG
SDP
Catalog No. 5000439137
Encompass_Preferred
8-CPT-Cyclic AMP sodium is a selective activator of cyclic AMP-dependent protein kinase (PKA) and a potent Epac activator. It also acts as an inhibitor for specific phosphodiesterases.
  • Selective activator of cyclic AMP-dependent protein kinase (PKA).
  • Potent inhibitor of cyclic GMP-specific phosphodiesterase (PDE VA) with an IC50 of 0.9 μM.
  • Inhibits PDE III and PDE IV.
  • Potent Epac activator.
Catalog No. 50-004-39137 Supplier Medchemexpress LLC Supplier No. HY11167350MG
8-CPT-Cyclic AMP sodium is a selective activator of cyclic AMP-dependent protein kinase (PKA) and a potent Epac activator. It also acts as an inhibitor for specific phosphodiesterases.
  • Selective activator of cyclic AMP-dependent protein kinase (PKA).
  • Potent inhibitor of cyclic GMP-specific phosphodiesterase (PDE VA) with an IC50 of 0.9 μM.
  • Inhibits PDE III and PDE IV.
  • Potent Epac activator.