Learn More
Medchemexpress LLC 8-Cyclopentyl-1,3-dimethylxanthine | 35873-49-5 | 99.8% | 248.28 | 5 MG
SDP
Catalog No. 5000452272
Encompass_Preferred
8-Cyclopentyl-1,3-dimethylxanthine is a selective adenosine A1 receptor antagonist with Ki values of 10.9 nM for the A1 receptor and 1440 nM for the A2 receptor. It is for research use only.
  • Selective adenosine A1 receptor antagonist
  • High purity (99.82%)
  • Available in solid and solution forms
Catalog No. 50-004-52272 Supplier Medchemexpress LLC Supplier No. HYW0119555MG
8-Cyclopentyl-1,3-dimethylxanthine is a selective adenosine A1 receptor antagonist with Ki values of 10.9 nM for the A1 receptor and 1440 nM for the A2 receptor. It is for research use only.
  • Selective adenosine A1 receptor antagonist
  • High purity (99.82%)
  • Available in solid and solution forms