Learn More
Medchemexpress LLC 8-Cyclopentyl-1,3-dipropylxanthine | 102146-07-6 | MFCD00055117 | ≥98.0% | 304.39 g/mol | C16H24N4O2 | 100 MG
SDP
Catalog No. 5000212942
Encompass_Preferred
DPCPX (PD 116948) is a xanthine-derived research compound that functions as a potent and selective adenosine A1 receptor antagonist. It is used in pharmacology and neuroscience research to block A1 receptor-mediated signaling in vitro and in vivo, supporting receptor binding, electrophysiology, and behavioral studies.
  • Potent and selective adenosine A1 receptor antagonist (Ki ≈ 0.46 nM).
  • Xanthine-derived compound for pharmacology and neuroscience research.
  • Suitable for receptor binding, electrophysiology, and behavioral assays.
  • Molecular formula C16H24N4O2; molecular weight 304.39 g/mol.
  • Typically soluble in DMSO for assay preparation.
Catalog No. 50-002-12942 Supplier Medchemexpress LLC Supplier No. HY100937100MG
DPCPX (PD 116948) is a xanthine-derived research compound that functions as a potent and selective adenosine A1 receptor antagonist. It is used in pharmacology and neuroscience research to block A1 receptor-mediated signaling in vitro and in vivo, supporting receptor binding, electrophysiology, and behavioral studies.
  • Potent and selective adenosine A1 receptor antagonist (Ki ≈ 0.46 nM).
  • Xanthine-derived compound for pharmacology and neuroscience research.
  • Suitable for receptor binding, electrophysiology, and behavioral assays.
  • Molecular formula C16H24N4O2; molecular weight 304.39 g/mol.
  • Typically soluble in DMSO for assay preparation.