Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 8-cyclopentyl-7-(cyclopentylmethyl)-2-(3,5-dichloro-4-hydroxyanilino)-5-methyl-7H-pteridin-6-one | 2411088-26-9 | 99.5% | C24H29Cl2N5O2 | 10MG
SDP

Catalog No. 5000261644
Encompass_Preferred

CC260 is a selective small-molecule inhibitor of PI5P4Kα and PI5P4Kβ used in research on cellular energy metabolism, diabetes, and cancer. The compound demonstrates low-nanomolar potency, high chemical purity, and is supplied with solubility and storage guidance suitable for in vitro and in vivo studies.

  • Selective inhibition of PI5P4Kα and PI5P4Kβ.
  • Potent binding (Ki ~40 nM and ~30 nM).
  • High purity (99.45%).
  • Soluble in DMSO at high concentration for in vitro use.
  • Formulation guidance provided for in vivo studies.
  • Recommended storage conditions for powder and solutions.

Catalog No. 50-002-61644 Supplier Medchemexpress LLC Supplier No. HY13918810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CC260 is a selective small-molecule inhibitor of PI5P4Kα and PI5P4Kβ used in research on cellular energy metabolism, diabetes, and cancer. The compound demonstrates low-nanomolar potency, high chemical purity, and is supplied with solubility and storage guidance suitable for in vitro and in vivo studies.

  • Selective inhibition of PI5P4Kα and PI5P4Kβ.
  • Potent binding (Ki ~40 nM and ~30 nM).
  • High purity (99.45%).
  • Soluble in DMSO at high concentration for in vitro use.
  • Formulation guidance provided for in vivo studies.
  • Recommended storage conditions for powder and solutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.