Learn More
Cayman Chemical 8-dehydro Cholesterol 10mg
SDP
Catalog No. 502936360
Encompass_Preferred
An isomer of 7-DHC; accumulates in tissues and plasma in patients with SLOS; levels are increased in serum in a rat model of SLOS induced by AY 9944
Catalog No. 50-293-6360 Supplier Cayman Chemical Supplier No. 2597010 MG
An isomer of 7-DHC; accumulates in tissues and plasma in patients with SLOS; levels are increased in serum in a rat model of SLOS induced by AY 9944