Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 8-dehydrocholesterol | 70741-38-7 | MFCD28359037 | 93.6% | 384.64 g·mol⁻¹ | C27H44O | 5 MG
SDP

Catalog No. 5000264181
Encompass_Preferred

8-Dehydrocholesterol is an endogenous cholesterol precursor whose elevated concentration is a diagnostic biochemical hallmark of classical Smith-Lemli-Opitz syndrome (SLOS). Supplied as a solid reference standard for research and analytical applications, it is characterized by documented chemical identifiers and supporting documentation for laboratory use.

  • CAS number 70741-38-7.
  • Purity 93.63% (analytical).
  • Molecular formula C27H44O; molecular weight 384.64 g·mol⁻¹.
  • Soluble in DMSO (5.56 mg/mL); ultrasonic warming to 60°C may improve solubility.
  • Store solid at -20°C, protect from light, and maintain under nitrogen; in solution store at -80°C up to 6 months.
  • Certificate of analysis and safety data sheet available for download.

Catalog No. 50-002-64181 Supplier Medchemexpress LLC Supplier No. HY1134355MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

8-Dehydrocholesterol is an endogenous cholesterol precursor whose elevated concentration is a diagnostic biochemical hallmark of classical Smith-Lemli-Opitz syndrome (SLOS). Supplied as a solid reference standard for research and analytical applications, it is characterized by documented chemical identifiers and supporting documentation for laboratory use.

  • CAS number 70741-38-7.
  • Purity 93.63% (analytical).
  • Molecular formula C27H44O; molecular weight 384.64 g·mol⁻¹.
  • Soluble in DMSO (5.56 mg/mL); ultrasonic warming to 60°C may improve solubility.
  • Store solid at -20°C, protect from light, and maintain under nitrogen; in solution store at -80°C up to 6 months.
  • Certificate of analysis and safety data sheet available for download.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.