Learn More
TARGETMOL CHEMICALS INC 8-Deoxygartanin 5MG
SDP
Catalog No. 502875819
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. 8-Deoxygartanin is a butyrylcholinesterase (BChE) selective inhibitor, it exhibits significant inhibition of self-induced alpha2-amyloid (Aalpha2) aggregation, it has multifunctional activities against Alzheimer's disease (AD) and could be promising compounds for the therapy of AD. Purity 99.76%
Catalog No. 50-287-5819 Supplier TARGETMOL CHEMICALS INC Supplier No. TN13415MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. 8-Deoxygartanin is a butyrylcholinesterase (BChE) selective inhibitor, it exhibits significant inhibition of self-induced alpha2-amyloid (Aalpha2) aggregation, it has multifunctional activities against Alzheimer's disease (AD) and could be promising compounds for the therapy of AD. Purity 99.76%