Learn More
Cayman Chemical 8-DesoxygartnIn 10mg
SDP
Catalog No. 502943592
Encompass_Preferred
A xanthone with anti-inflammatory and enzyme inhibitory activities; inhibits NF-κB p65 binding to DNA in a cell-free assay in an enzyme assay (IC50 = 11.3 µM); inhibits BChE and AChE (IC50s = 2.46 and 7.75 UG/ml)
Catalog No. 50-294-3592 Supplier Cayman Chemical Supplier No. 4027510 MG
A xanthone with anti-inflammatory and enzyme inhibitory activities; inhibits NF-κB p65 binding to DNA in a cell-free assay in an enzyme assay (IC50 = 11.3 µM); inhibits BChE and AChE (IC50s = 2.46 and 7.75 UG/ml)