Learn More
MSC 8 DIAL HANGING SCALE
Catalog No. 5000686204
Encompass
This hanging dial scale provides accurate measurements for various applications. It features a durable design with a plastic housing and steel components, and includes a galvanized scoop for convenience.
  • Hanging dial scale type
  • Features 8 in. dial display
  • Provides 1 oz. graduation
  • Offers 2 revolutions
  • Capacity of 10.00 lb. graduation
  • Plastic housing
  • Steel hook and frame
  • Galvanized scoop
  • Manufactured in the United States
Catalog No. 50-006-86204 Supplier MSC Supplier No. 65360752
This hanging dial scale provides accurate measurements for various applications. It features a durable design with a plastic housing and steel components, and includes a galvanized scoop for convenience.
  • Hanging dial scale type
  • Features 8 in. dial display
  • Provides 1 oz. graduation
  • Offers 2 revolutions
  • Capacity of 10.00 lb. graduation
  • Plastic housing
  • Steel hook and frame
  • Galvanized scoop
  • Manufactured in the United States