Learn More
Dual Manufacturing Company Inc 8'' Diameter x 2'' Deep Stainless Mesh Sieves
Catalog No. NC1820795
Encompass_Preferred
Stainless Steel. Mesh Openings: 710 Micron 0.710 mm 0.0278 in. Wire Diameter: 0.450 mm 0.017 in.
Catalog No. NC1820795 Supplier Dual Manufacturing Company Inc Supplier No. US825FS
Stainless Steel. Mesh Openings: 710 Micron 0.710 mm 0.0278 in. Wire Diameter: 0.450 mm 0.017 in.