Learn More
California Pacific Lab & Consulting 8" ECO FUNNEL,SYSTEM,10 LITER
SDP
Catalog No. NC1467988
Encompass_Preferred
8" ECO Funnel, System, 10 Liter, 83mm Cap adapter (83B)The 8 inch ECO Funnel with 10 Liter Waste Disposal System and 83mm screw cap features a latching lid with gasket to keep fumes contained. A removable large particle filter catches stir bars or other items. The 11" funnel stem reduces chemical evaporation, and the secondary air lock tube helps prevent over-filling.
Catalog No. NC1467988 Supplier California Pacific Lab & Consulting Supplier No. EF883CSYS
8" ECO Funnel, System, 10 Liter, 83mm Cap adapter (83B)The 8 inch ECO Funnel with 10 Liter Waste Disposal System and 83mm screw cap features a latching lid with gasket to keep fumes contained. A removable large particle filter catches stir bars or other items. The 11" funnel stem reduces chemical evaporation, and the secondary air lock tube helps prevent over-filling.