Learn More
California Pacific Lab & Consulting 8 ECO FUNNEL W/53MM SCREW CAP
SDP
Catalog No. NC0184260
Encompass_Preferred
8 in. SAFETY ECO FUNNEL FOR LIQUID WASTE COLLECTION, FITS 53MM SCREW CAP BOTTLES, PREVENTS 99.9% EMISSIONS OF TOXIC FUMES, HDPE, RED LID WITH GASKET THAT LATCHES, REMOVABLE FILTER FOR LARGE PARTICLES, 2000 ML CAPACITY, LIQUID LEVEL TUBE FOR PREVENTING OVERF
Catalog No. NC0184260 Supplier California Pacific Lab & Consulting Supplier No. EF3008
8 in. SAFETY ECO FUNNEL FOR LIQUID WASTE COLLECTION, FITS 53MM SCREW CAP BOTTLES, PREVENTS 99.9% EMISSIONS OF TOXIC FUMES, HDPE, RED LID WITH GASKET THAT LATCHES, REMOVABLE FILTER FOR LARGE PARTICLES, 2000 ML CAPACITY, LIQUID LEVEL TUBE FOR PREVENTING OVERF