Learn More
Medchemexpress LLC 8-Epidiosbulbin E acetate | 91095-48-6 | 50 MG
SDP
Catalog No. 5000418691
Encompass_Preferred
8-Epidiosbulbin E acetate is a furanoid found in Dioscorea bulbifera L. known for its broad-spectrum plasmid-curing activity against multidrug-resistant (MDR) bacteria. This compound is also noted for its ability to induce liver injury, making it useful in hepatotoxicity research.
  • Furanoid derived from Dioscorea bulbifera L.
  • Broad-spectrum plasmid-curing activity against multidrug-resistant (MDR) bacteria.
  • Induces liver injury, suitable for animal modeling and hepatotoxicity studies.
  • Hepatotoxicity mediated by metabolic activation of its furan ring by cytochromes P450.
  • Damages hepatic cells via aberrant regulation of bile acid, taurine, and hypotaurine metabolism.
  • For research use only.
  • Requires freshly opened DMSO for solubility; aliquot solutions to prevent inactivation.
  • Prepare working solutions freshly for in vivo experiments.
Catalog No. 50-004-18691 Supplier Medchemexpress LLC Supplier No. HYN704750MG
8-Epidiosbulbin E acetate is a furanoid found in Dioscorea bulbifera L. known for its broad-spectrum plasmid-curing activity against multidrug-resistant (MDR) bacteria. This compound is also noted for its ability to induce liver injury, making it useful in hepatotoxicity research.
  • Furanoid derived from Dioscorea bulbifera L.
  • Broad-spectrum plasmid-curing activity against multidrug-resistant (MDR) bacteria.
  • Induces liver injury, suitable for animal modeling and hepatotoxicity studies.
  • Hepatotoxicity mediated by metabolic activation of its furan ring by cytochromes P450.
  • Damages hepatic cells via aberrant regulation of bile acid, taurine, and hypotaurine metabolism.
  • For research use only.
  • Requires freshly opened DMSO for solubility; aliquot solutions to prevent inactivation.
  • Prepare working solutions freshly for in vivo experiments.