Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 8-Epiloganic acid | 82509-41-9 | 1 MG
SDP

Catalog No. 5000417449
Encompass_Preferred

8-Epiloganic acid is an iridoid glucoside, originating from plants such as Linaria cymbalaria (Scrophulariaceae). This compound is classified under terpenoids and iridoids.

  • Purity of 99.76%
  • Appears as a white to off-white solid
  • Store at 4°C, sealed, away from moisture and light
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month (sealed, away from moisture and light)

Catalog No. 50-004-17449 Supplier Medchemexpress LLC Supplier No. HYN83141MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

8-Epiloganic acid is an iridoid glucoside, originating from plants such as Linaria cymbalaria (Scrophulariaceae). This compound is classified under terpenoids and iridoids.

  • Purity of 99.76%
  • Appears as a white to off-white solid
  • Store at 4°C, sealed, away from moisture and light
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month (sealed, away from moisture and light)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.