Learn More
Medchemexpress LLC 8-Ethyl Irinotecan | 947687-02-7 | 25 MG
SDP
Catalog No. 5000341739
Encompass_Preferred
8-Ethyl Irinotecan (compound H) is an impurity of Irinotecan hydrochloride. It has a molecular weight of 614.73 and a chemical formula of C35H42N4O6. It appears as a solid with a light yellow to yellow color.
  • Target: Biochemical Assay Reagents
  • Pathway: Others
  • For research use only
  • Not fully validated for medical applications
Catalog No. 50-003-41739 Supplier Medchemexpress LLC Supplier No. 25MGHY131174
8-Ethyl Irinotecan (compound H) is an impurity of Irinotecan hydrochloride. It has a molecular weight of 614.73 and a chemical formula of C35H42N4O6. It appears as a solid with a light yellow to yellow color.
  • Target: Biochemical Assay Reagents
  • Pathway: Others
  • For research use only
  • Not fully validated for medical applications