Learn More
Medchemexpress LLC 8-ethyl irinotecan | 947687-02-7 | MFCD29920109 | ≥98.0% | C35H42N4O6 | 10MG
SDP
Catalog No. 5000261914
Encompass_Preferred
8-Ethyl irinotecan is an impurity of irinotecan hydrochloride used primarily as an analytical reference and research reagent. It is identified by CAS RN 947687-02-7, with molecular formula C35H42N4O6 and molecular weight 614.73 g/mol. The material is provided in small research quantities and users should verify lot-specific purity and storage instructions on the certificate of analysis.
  • Impurity of irinotecan hydrochloride useful for impurity profiling.
  • Suitable as an analytical reference standard in HPLC and mass spectrometry.
  • Molecular formula C35H42N4O6; molecular weight 614.73 g/mol.
  • Supplied in small quantities for research use; consult the certificate of analysis for purity.
  • Store according to the certificate of analysis to maintain stability.
Catalog No. 50-002-61914 Supplier Medchemexpress LLC Supplier No. HY13117410MG
8-Ethyl irinotecan is an impurity of irinotecan hydrochloride used primarily as an analytical reference and research reagent. It is identified by CAS RN 947687-02-7, with molecular formula C35H42N4O6 and molecular weight 614.73 g/mol. The material is provided in small research quantities and users should verify lot-specific purity and storage instructions on the certificate of analysis.
  • Impurity of irinotecan hydrochloride useful for impurity profiling.
  • Suitable as an analytical reference standard in HPLC and mass spectrometry.
  • Molecular formula C35H42N4O6; molecular weight 614.73 g/mol.
  • Supplied in small quantities for research use; consult the certificate of analysis for purity.
  • Store according to the certificate of analysis to maintain stability.