Learn More
Cayman Chemical 8-ethylthio NAD sodium salt
SDP
Catalog No. 502917241
Encompass_Preferred
A derivative of NAD+; has been used in the screening of analog-sensitive PARP1 gatekeeper mutations
Catalog No. 50-291-7241 Supplier Cayman Chemical Supplier No. 38483700 UG
A derivative of NAD+; has been used in the screening of analog-sensitive PARP1 gatekeeper mutations