Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 8-fluoro-5-methyl-6-oxo-4H-imidazo[1,5-a]benzodiazepine-3-carboxylic acid | 84378-44-9 | MFCD08061420 | 100.0% | 275.24 g/mol | C13H10FN3O3 | 25 MG
SDP

Catalog No. 5000229233
Encompass_Preferred

Flumazenil acid is the carboxylic acid metabolite of flumazenil (Ro 15-3890). Supplied as an off-white to light yellow solid reference compound for laboratory research, it is intended for analytical standards, metabolite studies, and assay development. The material is for research use only.

  • High purity, 99.96% as reported on supplier COA.
  • Molecular weight 275.24 g/mol.
  • Chemical formula C13H10FN3O3.
  • Appearance: off-white to light yellow solid.
  • Common application: metabolite standard and analytical reference.
  • Available unit: 25 MG typical pack size.

Catalog No. 50-002-29233 Supplier Medchemexpress LLC Supplier No. HY11884425MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Flumazenil acid is the carboxylic acid metabolite of flumazenil (Ro 15-3890). Supplied as an off-white to light yellow solid reference compound for laboratory research, it is intended for analytical standards, metabolite studies, and assay development. The material is for research use only.

  • High purity, 99.96% as reported on supplier COA.
  • Molecular weight 275.24 g/mol.
  • Chemical formula C13H10FN3O3.
  • Appearance: off-white to light yellow solid.
  • Common application: metabolite standard and analytical reference.
  • Available unit: 25 MG typical pack size.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.