Learn More
Alkali Scientific 8 Gallon Sharps Container, Hinged Lid, 10/Case
SDP
Catalog No. 5000662053
Encompass_Preferred
Alkali Scientific 8 Gallon Sharps Containers are packaged in cases of 10 units. The hinged cover with gasket provides leak resistant containment of fluids, and a pad at the bottom of each container absorbs any excess liquids. These sharps containers provide temporary closure when not in use and final locking closure for disposal. Large and small openings allow for disposal of various types of infectious waste and they easily accommodate the largest sharps, suction canisters, trocars, or endoscopic instruments. They are essential for any laboratory that works with biological samples. 10/Case 8 gallon capacity Hinged cover for temporary closure when not in use Easily accommodates the largest sharps Locks for final disposal Adjustable rotor opening Dimensions: 7.5”H x 11"D x 15.5"
Catalog No. 50-006-62053 Supplier Alkali Scientific Supplier No. KE8997
Alkali Scientific 8 Gallon Sharps Containers are packaged in cases of 10 units. The hinged cover with gasket provides leak resistant containment of fluids, and a pad at the bottom of each container absorbs any excess liquids. These sharps containers provide temporary closure when not in use and final locking closure for disposal. Large and small openings allow for disposal of various types of infectious waste and they easily accommodate the largest sharps, suction canisters, trocars, or endoscopic instruments. They are essential for any laboratory that works with biological samples. 10/Case 8 gallon capacity Hinged cover for temporary closure when not in use Easily accommodates the largest sharps Locks for final disposal Adjustable rotor opening Dimensions: 7.5”H x 11"D x 15.5"