Learn More
Medchemexpress LLC 8-Geranyloxypsoralen | 7437-55-0 | MFCD03412052 | 99.9% | 338.40 g/mol | C21H22O4 | 10 MG
SDP
Catalog No. 5000289374
Encompass_Preferred
8-Geranyloxypsoralen is a furanocoumarin compound isolated from grapefruit that functions as a cytochrome P450 3A4 (CYP3A4) inhibitor. It is provided for biochemical and pharmacological research to study CYP-mediated metabolism and potential drug-drug interactions.
  • Potent CYP3A4 inhibitor with reported IC50 of 3.93 μM.
  • High purity (99.9%).
  • Molecular formula C21H22O4; molecular weight 338.40 g/mol.
  • Supplied in small-scale quantities suitable for research, including 10 mg packs.
  • Applicable to in vitro enzyme assays and metabolism studies.
Catalog No. 50-002-89374 Supplier Medchemexpress LLC Supplier No. HYN226210MG
8-Geranyloxypsoralen is a furanocoumarin compound isolated from grapefruit that functions as a cytochrome P450 3A4 (CYP3A4) inhibitor. It is provided for biochemical and pharmacological research to study CYP-mediated metabolism and potential drug-drug interactions.
  • Potent CYP3A4 inhibitor with reported IC50 of 3.93 μM.
  • High purity (99.9%).
  • Molecular formula C21H22O4; molecular weight 338.40 g/mol.
  • Supplied in small-scale quantities suitable for research, including 10 mg packs.
  • Applicable to in vitro enzyme assays and metabolism studies.