Learn More
Medchemexpress LLC HY-N0447 5mg Medchemexpress, 8-Gingerol CAS:23513-08-8 Purity:>98%
SDP
Catalog No. 501970271
Encompass_Preferred
Medchemexpress, HY-N0447 5mg 8-Gingerol CAS:23513-08-8 8-Gingerol, found in the rhizomes of ginger (Z. officinale) with oral bioavailability, activates TRPV1, with an EC50 of 5.0 µM. 8-Gingerol inhibits COX-2, and inhibits the growth of H. pylori in vitro. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0271 Supplier Medchemexpress LLC Supplier No. HYN04475MG
Medchemexpress, HY-N0447 5mg 8-Gingerol CAS:23513-08-8 8-Gingerol, found in the rhizomes of ginger (Z. officinale) with oral bioavailability, activates TRPV1, with an EC50 of 5.0 µM. 8-Gingerol inhibits COX-2, and inhibits the growth of H. pylori in vitro. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.