Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[8]-Gingerol, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013629 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3629 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3629 Supplier MilliporeSigma Supelco Supplier No. 0151410MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Gingerol is the pungent component of ginger roots. 8-Gingerol has a chain length of n8. It is a homologous series of phenolic ketones.

Featured Industries: Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

CAS 23513-08-8
Linear Formula C19H30O4
MDL Number MFCD00877794
Synonym (S)-5-Hydroxy-1-(4-hydroxy-3-methoxyphenyl)-3-dodecanone; 8-Gingerol
Type Analytical Standard
Quantity 10 mg
Formula Weight 322.44
Percent Purity ≥95% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.