Learn More
Pipette Supplies, Inc. Eppendorf Silicone Grease with Lint-Free Applicators
SDP
Catalog No. NC1868591
Encompass
Pipette grease, 8 grams, with stick swabs. Eppendorf silicone grease should be used to grease pistons when pipette has become extremely dirty and must be cleaned with organic solvents. Apply grease following cleaning.
Catalog No. NC1868591 Supplier Pipette Supplies, Inc. Supplier No. EP 0013022153
Pipette grease, 8 grams, with stick swabs. Eppendorf silicone grease should be used to grease pistons when pipette has become extremely dirty and must be cleaned with organic solvents. Apply grease following cleaning.