Learn More
Medchemexpress LLC HY-W011540 10mg Medchemexpress, 8-Hydroxy-2'-deoxyguanosine CAS:88847-89-6 Purity:>98%
SDP
Catalog No. 501934552
Encompass_Preferred
Medchemexpress, HY-W011540 10mg 8-Hydroxy-2'-deoxyguanosine CAS:88847-89-6 8-Hydroxy-2'-deoxyguanosine is a critical biomarker of oxidative stress and carcinogenesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4552 Supplier Medchemexpress LLC Supplier No. HYW01154010MG
Medchemexpress, HY-W011540 10mg 8-Hydroxy-2'-deoxyguanosine CAS:88847-89-6 8-Hydroxy-2'-deoxyguanosine is a critical biomarker of oxidative stress and carcinogenesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.