Learn More
TARGETMOL CHEMICALS INC 8-Hydroxy-2-deoxyguanosine 2MG
SDP
Catalog No. 502876716
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. 8-Hydroxy-2'-deoxyguanosine (8-OHdG) is a good biomarker for risk assessment of various cancers and degenerative diseases. Purity 97.13%
Catalog No. 50-287-6716 Supplier TARGETMOL CHEMICALS INC Supplier No. T83432MG
Also available in 1 mL, 1 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. 8-Hydroxy-2'-deoxyguanosine (8-OHdG) is a good biomarker for risk assessment of various cancers and degenerative diseases. Purity 97.13%