Learn More
Abcam 8-hydroxy 2 deoxyguanosine ELISA Kit, 1 x 96 Tests
Catalog No. NC1328263
Encompass_Preferred
The 8-Hydroxy-2'-deoxyguanosine (8-OHdG) ELISA kit is a competitive ELISA designed to quantify 8-Hydroxy-2'-deoxyguanosine with a sensitivity of 0.59 ng/mL. - Marker for oxidative DNA damage - Wide dynamic range – quantifies 0.94–60 ng/mL - Compatible with multiple sample types – works with serum, plasma, saliva, urine, cell lysates, and tissue extracts - Cited in over 100 publications
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1328263 Supplier Abcam Supplier No. AB2017341X96TESTS
Add to Cart
Add to Cart
The 8-Hydroxy-2'-deoxyguanosine (8-OHdG) ELISA kit is a competitive ELISA designed to quantify 8-Hydroxy-2'-deoxyguanosine with a sensitivity of 0.59 ng/mL. - Marker for oxidative DNA damage - Wide dynamic range – quantifies 0.94–60 ng/mL - Compatible with multiple sample types – works with serum, plasma, saliva, urine, cell lysates, and tissue extracts - Cited in over 100 publications
The product is subject to the following: Abcam Restricted Use Statement