Learn More
Abcam 8-Hydro x y-2'-deo x yguanosine, O x idative stress biomarker, 5MG
Catalog No. 502705125
Encompass_Preferred
MW 283.24 Da, Purity >98%. Oxidative stress biomarker. A predominant form of free radical-induced oxidative lesions in nuclear and mitochondrial DNA. Oxidized deoxyguanosine derivative. Biomarker for oxidative stress and carcinogenesis.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5125 Supplier Abcam Supplier No. AB1455955MG
MW 283.24 Da, Purity >98%. Oxidative stress biomarker. A predominant form of free radical-induced oxidative lesions in nuclear and mitochondrial DNA. Oxidized deoxyguanosine derivative. Biomarker for oxidative stress and carcinogenesis.
The product is subject to the following: Abcam Restricted Use Statement