Learn More
Cayman Chemical 8-Hydroxy-2-deoxygunosIn 10mg
SDP
Catalog No. 502896327
Encompass_Preferred
8-Hydroxy-2'-deoxyguanosine is produced by oxidative damage of DNA by reactive oxygen and nitrogen species, including hydroxyl radical and peroxynitrite. It serves as a marker of oxidative stress in biological systems.{1239,5022,2239,3718}
Catalog No. 50-289-6327 Supplier Cayman Chemical Supplier No. 8932010 MG
8-Hydroxy-2'-deoxyguanosine is produced by oxidative damage of DNA by reactive oxygen and nitrogen species, including hydroxyl radical and peroxynitrite. It serves as a marker of oxidative stress in biological systems.{1239,5022,2239,3718}