Learn More
Medchemexpress LLC 8-hydroxy-DPAT hydrobromide | 76135-31-4 | MFCD00055158 | 99.9% | 328.29 | C16H26BrNO | 10 MG
SDP
Catalog No. 5000287736
Encompass_Preferred
8-hydroxy-DPAT hydrobromide is the hydrobromide salt of 8-hydroxy-2-dipropylaminotetralin, a potent and selective agonist of the serotonin 5-HT1A receptor used in neuropharmacology research. Supplied as a high-purity solid, it is suitable for in vitro and in vivo studies and is provided with analytical documentation.
  • Potent, selective 5-HT1A receptor agonist (pIC50 ~8.19).
  • Hydrobromide salt for improved stability and handling.
  • High purity (>99.8%) suitable for research applications.
  • Available as a solid; common pack size 10 mg.
  • Provided with COA, 1H NMR, and LC-MS documentation.
  • Molecular formula C16H26BrNO; molecular weight 328.29.
Catalog No. 50-002-87736 Supplier Medchemexpress LLC Supplier No. HY1568810MG
8-hydroxy-DPAT hydrobromide is the hydrobromide salt of 8-hydroxy-2-dipropylaminotetralin, a potent and selective agonist of the serotonin 5-HT1A receptor used in neuropharmacology research. Supplied as a high-purity solid, it is suitable for in vitro and in vivo studies and is provided with analytical documentation.
  • Potent, selective 5-HT1A receptor agonist (pIC50 ~8.19).
  • Hydrobromide salt for improved stability and handling.
  • High purity (>99.8%) suitable for research applications.
  • Available as a solid; common pack size 10 mg.
  • Provided with COA, 1H NMR, and LC-MS documentation.
  • Molecular formula C16H26BrNO; molecular weight 328.29.