Learn More
Medchemexpress LLC 8-Hydroxyguanosine | 3868-31-3 | 99.9% | 299.24 | 100 MG
SDP
Catalog No. 5000437400
Encompass_Preferred
8-Hydroxyguanosine is an oxidized nucleoside, a marker of RNA oxidative damage and oxidative stress. It stimulates proliferation and differentiation of murine B cells, exhibiting immunostimulatory activity. This compound is promising for research of Alzheimer's disease and Down's syndrome.
  • Marker of RNA oxidative damage and oxidative stress
  • Stimulates proliferation and differentiation of murine B cells
  • Exhibits immunostimulatory activity
  • Promising for research of Alzheimer's disease and Down's syndrome
Catalog No. 50-004-37400 Supplier Medchemexpress LLC Supplier No. HY113262100MG
8-Hydroxyguanosine is an oxidized nucleoside, a marker of RNA oxidative damage and oxidative stress. It stimulates proliferation and differentiation of murine B cells, exhibiting immunostimulatory activity. This compound is promising for research of Alzheimer's disease and Down's syndrome.
  • Marker of RNA oxidative damage and oxidative stress
  • Stimulates proliferation and differentiation of murine B cells
  • Exhibits immunostimulatory activity
  • Promising for research of Alzheimer's disease and Down's syndrome