Learn More
Abcam 8-Hydro x yguanosine, biomarker for o x idative damage, 1MG
Catalog No. 502705127
Encompass_Preferred
MW 299.24 Da, Purity >98%. Biomarker for oxidative damage to nucleic acids and lipids. Product of oxidative damage of RNA. Promotes differentiation and shows antiproliferative effects.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5127 Supplier Abcam Supplier No. AB1455941MG
MW 299.24 Da, Purity >98%. Biomarker for oxidative damage to nucleic acids and lipids. Product of oxidative damage of RNA. Promotes differentiation and shows antiproliferative effects.
The product is subject to the following: Abcam Restricted Use Statement