Learn More
Enzo Life Sciences 8-Hydroxyguanosine polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences ALX210802R100
Encompass_Preferred
Host: Goat. Immunogen: Synthetic 8-hydroxyguanosine-conjugate. Species reactivity: Species independent. Specificity: Recognizes 8-hydroxyguanosine and 8-hydroxy-2'-deoxyguanosine. Applications: ELISA, IHC (FS), IHC (PS). Formulation: Liquid. Neat serum. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-8160