Learn More
Cayman Chemical 8-HydroxygunInhydrochlorI 50mg
SDP
Catalog No. 502894040
Encompass_Preferred
A polyamine with diverse biological activities scavenges DPPH superoxide and hydroxyl radicals in cell-free assays (IC50s 38.5 291.6 and 351.9 UM respectively) inhibits tyrosinase (IC50 291.3 UM for the mushroom enzyme) and forskolin-induced pigmentation in B16/F10 cells at 50 UG/ml inhibits LPS-induced increases in NO and iNOS levels as well as NF-KB activity in RAW 264.7 macrophages at 50 UM
Catalog No. 50-289-4040 Supplier Cayman Chemical Supplier No. 8929050 MG
A polyamine with diverse biological activities scavenges DPPH superoxide and hydroxyl radicals in cell-free assays (IC50s 38.5 291.6 and 351.9 UM respectively) inhibits tyrosinase (IC50 291.3 UM for the mushroom enzyme) and forskolin-induced pigmentation in B16/F10 cells at 50 UG/ml inhibits LPS-induced increases in NO and iNOS levels as well as NF-KB activity in RAW 264.7 macrophages at 50 UM