Learn More
AdipoGen 8-Hydroxyjulolidine
SDP
Supplier:  AdipoGen CDXH0025G001
Encompass_Preferred
Chemical. CAS 41175-50-2. Formula C12H15NO. MW 189.25. Synthetic. An intermediate and starting material for various coumarin syntheses. Used to prepare aminocoumarins via microwave-accelerated Pechmann reaction or as starting material for coumarin C-ribosides.
Catalog No. 50-256-0799