Learn More
Medchemexpress LLC 8-Hydroxyquinoline | 10 MM 1 ML
SDP
Catalog No. 5000407524
Encompass_Preferred
8-Hydroxyquinoline (8-Quinolinol) is a versatile compound acting as a lipophilic metal chelator and a fungicide. It demonstrates activity against *Neisseria gonorrhoeae* and can interact with copper to facilitate cellular transport. Research indicates its potential to increase micronucleated polychromatic erythrocytes and cause hair depigmentation in mice.
  • Acts as a lipophilic metal chelator
  • Functions as a fungicide
  • Effective against *Neisseria gonorrhoeae*
  • Binds to copper and facilitates transport into cells
  • Increases micronucleated polychromatic erythrocytes
  • Can cause hair depigmentation in mice
Catalog No. 50-004-07524 Supplier Medchemexpress LLC Supplier No. HYB100510MM/1ML
8-Hydroxyquinoline (8-Quinolinol) is a versatile compound acting as a lipophilic metal chelator and a fungicide. It demonstrates activity against *Neisseria gonorrhoeae* and can interact with copper to facilitate cellular transport. Research indicates its potential to increase micronucleated polychromatic erythrocytes and cause hair depigmentation in mice.
  • Acts as a lipophilic metal chelator
  • Functions as a fungicide
  • Effective against *Neisseria gonorrhoeae*
  • Binds to copper and facilitates transport into cells
  • Increases micronucleated polychromatic erythrocytes
  • Can cause hair depigmentation in mice