Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Hydroxyquinoline-7-carboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. H08775G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08775G 5 g
H087725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H08775G Supplier TCI America Supplier No. H08775G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 19829-79-9
Molecular Formula C10H7NO3
Molecular Weight (g/mol) 189.17
MDL Number MFCD00671539
InChI Key JYIAZVFJRYLCBH-UHFFFAOYSA-N
PubChem CID 231363
IUPAC Name 8-hydroxyquinoline-7-carboxylic acid
SMILES C1=CC2=C(C(=C(C=C2)C(=O)O)O)N=C1

Specifications

Quantity 5 g
Formula Weight 189.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 8-Hydroxyquinoline-7-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.