Learn More
8-Hydroxyquinoline, ACS reagent, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
Specifications
Specifications
| Melting Point | 72.0°C to 74.0°C |
| Color | Beige or White to Yellow |
| Boiling Point | 267.0°C (752.0 mmHg) |
| Ignition Residue | 0.05% max. |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99% |
| Packaging | Glass bottle |
| Quantity | 100 g |
| Beilstein | 21, IV, 1135 |
| Merck Index | 15, 4881 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Toxic if swallowed. May cause an allergic skin reaction. Causes serious eye damage. May damage fertility or the unborn child. Very toxic to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doc |
| EINECSNumber | 205-711-1 |
| RTECSNumber | VC4200000 |
| TSCA | TSCA |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.