Learn More
Medchemexpress LLC 8-Hydroxyquinoline hemisulfate | 134-31-6 | 99.6% | 194.20 | 50 MG
SDP
Catalog No. 5000452787
Encompass_Preferred
8-Hydroxyquinoline (hemisulfate) (Standard) serves as the analytical standard for 8-Hydroxyquinoline (hemisulfate), and is designated for research and analytical applications. It is recognized as a monoprotic bidentate chelating agent that possesses antiseptic, disinfectant, and pesticide properties, and also functions as a transcription inhibitor.
  • Analytical standard for 8-Hydroxyquinoline (hemisulfate).
  • Intended for research and analytical applications.
  • Functions as a monoprotic bidentate chelating agent.
  • Exhibits antiseptic, disinfectant, and pesticide properties.
  • Acts as a transcription inhibitor.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-004-52787 Supplier Medchemexpress LLC Supplier No. HYW012037R50MG
8-Hydroxyquinoline (hemisulfate) (Standard) serves as the analytical standard for 8-Hydroxyquinoline (hemisulfate), and is designated for research and analytical applications. It is recognized as a monoprotic bidentate chelating agent that possesses antiseptic, disinfectant, and pesticide properties, and also functions as a transcription inhibitor.
  • Analytical standard for 8-Hydroxyquinoline (hemisulfate).
  • Intended for research and analytical applications.
  • Functions as a monoprotic bidentate chelating agent.
  • Exhibits antiseptic, disinfectant, and pesticide properties.
  • Acts as a transcription inhibitor.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.