Learn More
Medchemexpress LLC 8-Hydroxyquinoline hemisulfate | 134-31-6 | 99.63% | 194.20 | 100 MG
SDP
Catalog No. 5000452985
Encompass_Preferred
8-Hydroxyquinoline (hemisulfate) (Standard) is the analytical standard of 8-Hydroxyquinoline (hemisulfate). This product is intended for research and analytical applications. 8-Hydroxyquinoline hemisulfate (8-Quinolinol hemisulfate) is a monoprotic bidentate chelating agent, exhibits antiseptic, disinfectant, and pesticide properties, functioning as a transcription inhibitor.
  • Monoprotic bidentate chelating agent
  • Exhibits antiseptic properties
  • Exhibits disinfectant properties
  • Exhibits pesticide properties
  • Functions as a transcription inhibitor
  • The grade of analytical standard, which is the reference standard supplied assay
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS
Catalog No. 50-004-52985 Supplier Medchemexpress LLC Supplier No. HYW012037R100MG
8-Hydroxyquinoline (hemisulfate) (Standard) is the analytical standard of 8-Hydroxyquinoline (hemisulfate). This product is intended for research and analytical applications. 8-Hydroxyquinoline hemisulfate (8-Quinolinol hemisulfate) is a monoprotic bidentate chelating agent, exhibits antiseptic, disinfectant, and pesticide properties, functioning as a transcription inhibitor.
  • Monoprotic bidentate chelating agent
  • Exhibits antiseptic properties
  • Exhibits disinfectant properties
  • Exhibits pesticide properties
  • Functions as a transcription inhibitor
  • The grade of analytical standard, which is the reference standard supplied assay
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS