Learn More
Grainger 8-Inch Work Boot M 8 Brown PR
Catalog No. 502768553
Encompass_Preferred
The Grainger 16P488 is a 3M Scotch-Brite General Purpose Hand Pad that can be used for cleaning and scouring. It features an open web construction with non-scratching fibers that are ideal for cleaning pots, pans, dishes, countertops, sinks, showers, tubs, fiberglass, porcelain, stainless steel and other surfaces. This 6 inch by 9 inch hand pad has a green color and comes in a pack of 10. It is durable, disposable and can remove grease, grime, food residue and more without scratching.
Catalog No. 50-276-8553 Supplier Grainger Supplier No. 16P488
The Grainger 16P488 is a 3M Scotch-Brite General Purpose Hand Pad that can be used for cleaning and scouring. It features an open web construction with non-scratching fibers that are ideal for cleaning pots, pans, dishes, countertops, sinks, showers, tubs, fiberglass, porcelain, stainless steel and other surfaces. This 6 inch by 9 inch hand pad has a green color and comes in a pack of 10. It is durable, disposable and can remove grease, grime, food residue and more without scratching.