Learn More
Grainger 8-Inch Work Boot M 9 1/2 Brown PR
Catalog No. 502765750
Encompass_Preferred
The 8-Inch Work Boot M 9-1/2 Brown is a durable work boot made by Wolverine. It features a full-grain leather upper, direct-attach PU midsole, and rubber lug outsole. The boot has a steel shank for arch support and stability. It has a removable full-cushion footbed for comfort. The boot meets ASTM F2413-18 M I/75 C/75 standards for impact and compression resistance. It is electrical hazard rated. The boot is designed for industrial work environments.
Catalog No. 50-276-5750 Supplier Grainger Supplier No. 16P494
The 8-Inch Work Boot M 9-1/2 Brown is a durable work boot made by Wolverine. It features a full-grain leather upper, direct-attach PU midsole, and rubber lug outsole. The boot has a steel shank for arch support and stability. It has a removable full-cushion footbed for comfort. The boot meets ASTM F2413-18 M I/75 C/75 standards for impact and compression resistance. It is electrical hazard rated. The boot is designed for industrial work environments.