Learn More
Grainger 8-Inch Work Boot W 10 1/2 Brown PR
Catalog No. 502765669
Encompass_Preferred
The Grainger 16P499 is a 16-ounce can of Rust-Oleum Stops Rust Protective Enamel spray paint in Gloss Black color. This oil-based paint provides a durable protective coating that resists rust, corrosion, fading and chipping. It features a comfort spray tip with a wider finger pad to reduce fatigue caused by continuous spraying. The advanced spray system allows paint to spray at any angle, even upside down, for full coverage. It is ideal for use on metal, wood, concrete and more. This enamel paint dries to the touch in 20 minutes and covers up to 15 sq ft.
Catalog No. 50-276-5669 Supplier Grainger Supplier No. 16P499
The Grainger 16P499 is a 16-ounce can of Rust-Oleum Stops Rust Protective Enamel spray paint in Gloss Black color. This oil-based paint provides a durable protective coating that resists rust, corrosion, fading and chipping. It features a comfort spray tip with a wider finger pad to reduce fatigue caused by continuous spraying. The advanced spray system allows paint to spray at any angle, even upside down, for full coverage. It is ideal for use on metal, wood, concrete and more. This enamel paint dries to the touch in 20 minutes and covers up to 15 sq ft.