Learn More
Grainger 8-Inch Work Boot W 8 1/2 Brown PR
Catalog No. 502767198
Encompass_Preferred
The Grainger 16P491 is a 1/2 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is totally enclosed, fan cooled (TEFC) with a cast iron body, aluminum end brackets, and double sealed ball bearings. It can be used for general purpose applications and is ideal for pumps, fans, conveyors, and other industrial machinery. This electric motor would be a good choice when a rugged, dependable 1/2 HP motor is needed.
Catalog No. 50-276-7198 Supplier Grainger Supplier No. 16P491
The Grainger 16P491 is a 1/2 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is totally enclosed, fan cooled (TEFC) with a cast iron body, aluminum end brackets, and double sealed ball bearings. It can be used for general purpose applications and is ideal for pumps, fans, conveyors, and other industrial machinery. This electric motor would be a good choice when a rugged, dependable 1/2 HP motor is needed.