Learn More
Grainger 8-Inch Work Boot W 8 Brown PR
Catalog No. 502767338
Encompass_Preferred
The Grainger 16P489 is a 16-ounce can of Rust-Oleum Stops Rust Protective Enamel spray paint in Gloss Black color. This oil-based paint provides a durable protective coating that resists corrosion, fading, chipping, and abrasion. It features a comfort spray tip with a wider finger pad to reduce fatigue caused by continuous spraying. The advanced spray system allows paint to spray at any angle, even upside down, for full coverage. It is ideal for coating metal, wood, plastic, wicker, and more. This enamel spray paint dries to the touch in 20 minutes and covers up to 15 sq ft per can.
Catalog No. 50-276-7338 Supplier Grainger Supplier No. 16P489
The Grainger 16P489 is a 16-ounce can of Rust-Oleum Stops Rust Protective Enamel spray paint in Gloss Black color. This oil-based paint provides a durable protective coating that resists corrosion, fading, chipping, and abrasion. It features a comfort spray tip with a wider finger pad to reduce fatigue caused by continuous spraying. The advanced spray system allows paint to spray at any angle, even upside down, for full coverage. It is ideal for coating metal, wood, plastic, wicker, and more. This enamel spray paint dries to the touch in 20 minutes and covers up to 15 sq ft per can.