Learn More
Grainger 8-Inch Work Boot W 9 1/2 Brown PR
Catalog No. 502767399
Encompass_Preferred
The 16P495 is a 16-ounce can of Rust-Oleum Stops Rust Protective Enamel spray paint in Gloss Black color. This oil-based paint provides a durable protective coating that resists rust, corrosion, fading, chipping, and abrasion. It features a comfort spray tip with a wider finger pad to reduce fatigue caused by continuous spraying. The advanced spray system allows you to spray at any angle, even upside down. It dries to the touch in 2 hours and covers up to 15 sq ft. This enamel spray paint is ideal for use on metal, wood, plastic, wicker and more.
Catalog No. 50-276-7399 Supplier Grainger Supplier No. 16P495
The 16P495 is a 16-ounce can of Rust-Oleum Stops Rust Protective Enamel spray paint in Gloss Black color. This oil-based paint provides a durable protective coating that resists rust, corrosion, fading, chipping, and abrasion. It features a comfort spray tip with a wider finger pad to reduce fatigue caused by continuous spraying. The advanced spray system allows you to spray at any angle, even upside down. It dries to the touch in 2 hours and covers up to 15 sq ft. This enamel spray paint is ideal for use on metal, wood, plastic, wicker and more.