Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Iodoquinoline 98.0+%, TCI America™
SDP

Catalog No. I0965200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0965200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I0965200MG Supplier TCI America Supplier No. I0965200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1006-47-9
Molecular Formula C9H6IN
Molecular Weight (g/mol) 255.058
MDL Number MFCD08062396
InChI Key OBEGSKDGFBFMFJ-UHFFFAOYSA-N
PubChem CID 17750386
IUPAC Name 8-iodoquinoline
SMILES C1=CC2=C(C(=C1)I)N=CC=C2

Specifications

Melting Point 33°C
Quantity 200 mg
Formula Weight 255.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 8-Iodoquinoline
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.